Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAAGCAGCACCTTGTTTCTTATTC[G/A]ATCTCTCTTGACCACTTCTTCTTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300298 | ||||||||||||||||||||
Literature Links: |
UPF3B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
UPF3B - UPF3 regulator of nonsense transcripts homolog B (yeast) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_023010.3 | 1388 | Silent Mutation | NP_075386.1 | |||
NM_080632.2 | 1388 | Silent Mutation | NP_542199.1 | |||
XM_017029737.1 | 1388 | Nonsense Mutation | XP_016885226.1 | |||
XM_017029738.1 | 1388 | Silent Mutation | XP_016885227.1 | |||
XM_017029739.1 | 1388 | Nonsense Mutation | XP_016885228.1 | |||
XM_017029740.1 | 1388 | Nonsense Mutation | XP_016885229.1 |