Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCTTTGGTTTCTGAGGCCTCCTG[A/G]GCCCTTCTGTACTGGGAGACCGCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300820 MIM: 300502 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAP3K15 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAP3K15 - mitogen-activated protein kinase kinase kinase 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001671.3 | 2561 | Silent Mutation | GCC,GCT | A,A 1303 | NP_001001671.3 | |
XM_011545507.2 | 2561 | Silent Mutation | GCC,GCT | A,A 1314 | XP_011543809.2 | |
XM_011545508.2 | 2561 | Silent Mutation | GCC,GCT | A,A 1285 | XP_011543810.2 | |
XM_011545510.2 | 2561 | Silent Mutation | GCC,GCT | A,A 861 | XP_011543812.1 | |
XM_011545511.2 | 2561 | Silent Mutation | GCC,GCT | A,A 738 | XP_011543813.1 | |
XM_017029511.1 | 2561 | Intron | XP_016885000.1 |
PDHA1 - pyruvate dehydrogenase (lipoamide) alpha 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000284.3 | 2561 | UTR 3 | NP_000275.1 | |||
NM_001173454.1 | 2561 | UTR 3 | NP_001166925.1 | |||
NM_001173455.1 | 2561 | UTR 3 | NP_001166926.1 | |||
NM_001173456.1 | 2561 | UTR 3 | NP_001166927.1 | |||
XM_017029574.1 | 2561 | Intron | XP_016885063.1 |