Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGAGGCAGCAGAAGCCGATCTAC[A/G]CCAGGGAGCGGGACCGGAAGCTGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 108728 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACLY PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACLY - ATP citrate lyase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TTC25 - tetratricopeptide repeat domain 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031421.3 | 840 | Missense Mutation | ACC,GCC | T,A 247 | NP_113609.1 | |
XM_011525323.2 | 840 | Missense Mutation | ACC,GCC | T,A 84 | XP_011523625.1 | |
XM_011525324.2 | 840 | Missense Mutation | ACC,GCC | T,A 247 | XP_011523626.1 |