Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGGGGAGGCGCGCCACTGGCTCA[A/C]GCAGTTCCAGACCTGCCATCACTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608300 MIM: 600781 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NAGS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NAGS - N-acetylglutamate synthase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153006.2 | 42 | Missense Mutation | AAG,ACG | K,T 125 | NP_694551.1 | |
XM_011524438.1 | 42 | Missense Mutation | AAG,ACG | K,T 125 | XP_011522740.1 | |
XM_011524439.2 | 42 | UTR 5 | XP_011522741.1 |
PYY - peptide YY | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004160.5 | 42 | Intron | NP_004151.3 | |||
XM_011525035.1 | 42 | Intron | XP_011523337.1 |
TMEM101 - transmembrane protein 101 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |