Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGAGGGCTGCAGCTGCGAACGAAC[G/T]CCCGCGCCACCCAGGGCCGGCGCCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606614 | ||||||||||||||||||||
Literature Links: |
RASGRF2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RASGRF2 - Ras protein specific guanine nucleotide releasing factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006909.2 | 278 | Silent Mutation | ACG,ACT | T,T 76 | NP_008840.1 | |
XM_005248565.1 | 278 | Silent Mutation | ACG,ACT | T,T 76 | XP_005248622.1 | |
XM_017009682.1 | 278 | Intron | XP_016865171.1 | |||
XM_017009683.1 | 278 | Silent Mutation | ACG,ACT | T,T 76 | XP_016865172.1 |
RASGRF2-AS1 - RASGRF2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |