Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGCATGTTGACAATCCTCACACA[A/G]CCTGCACAGGGAAGGAATTGGTGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603166 | ||||||||||||||||||||
Literature Links: |
MAP4K2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MAP4K2 - mitogen-activated protein kinase kinase kinase kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001307990.1 | 2106 | Silent Mutation | CGC,CGT | R,R 713 | NP_001294919.1 | |
NM_004579.4 | 2106 | Silent Mutation | CGC,CGT | R,R 721 | NP_004570.2 | |
XM_011545203.2 | 2106 | Silent Mutation | GGC,GGT | G,G 698 | XP_011543505.1 | |
XM_011545204.2 | 2106 | Silent Mutation | CGC,CGT | R,R 677 | XP_011543506.1 | |
XM_017018093.1 | 2106 | Silent Mutation | CGC,CGT | R,R 669 | XP_016873582.1 | |
XM_017018094.1 | 2106 | Silent Mutation | CGC,CGT | R,R 544 | XP_016873583.1 | |
XM_017018095.1 | 2106 | Silent Mutation | GGC,GGT | G,G 521 | XP_016873584.1 | |
XM_017018096.1 | 2106 | Intron | XP_016873585.1 |