Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGACTGGGGGCCCCATGGAAAGCC[A/G]CAGGGTGCAGGAGGTAGGCCTCAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602035 MIM: 602427 MIM: 609724 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101928595 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC101928595 - uncharacterized LOC101928595 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPP4C - protein phosphatase 4 catalytic subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TBX6 - T-box 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004608.3 | 1146 | Missense Mutation | GCG,GTG | A,V 354 | NP_004599.2 | |
XM_005255523.2 | 1146 | Missense Mutation | GCG,GTG | A,V 354 | XP_005255580.1 | |
XM_011545926.2 | 1146 | Missense Mutation | GCG,GTG | A,V 354 | XP_011544228.1 | |
XM_017023614.1 | 1146 | Missense Mutation | GCG,GTG | A,V 354 | XP_016879103.1 |
YPEL3 - yippee like 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |