Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACGAGGAAGACGCCGAGGCCTGGG[C/G]CATGGAACTGGCGGACGTGGGGGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605252 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
POLI PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
POLI - DNA polymerase iota | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007195.2 | 63 | Missense Mutation | GCC,GGC | A,G 25 | NP_009126.2 | |
XM_005258188.3 | 63 | UTR 5 | XP_005258245.1 | |||
XM_005258192.4 | 63 | Missense Mutation | GCC,GGC | A,G 25 | XP_005258249.1 | |
XM_006722378.2 | 63 | Missense Mutation | GCC,GGC | A,G 25 | XP_006722441.1 | |
XM_011525796.2 | 63 | UTR 5 | XP_011524098.1 | |||
XM_011525797.1 | 63 | Missense Mutation | GCC,GGC | A,G 25 | XP_011524099.1 |