Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATCGAGGGTATAAGCACACGTCT[C/T]GGATGTGATACCTATTCAGAATCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 108410 | ||||||||||||||||||||
Literature Links: |
NARS PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NARS - asparaginyl-tRNA synthetase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004539.3 | 1829 | Missense Mutation | CAA,CGA | Q,R 534 | NP_004530.1 | |
XM_005266700.1 | 1829 | Missense Mutation | CAA,CGA | Q,R 533 | XP_005266757.1 |