Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
CACGTACTCGCGTGCCCGCAGTGGC[A/G]AGCGGTTCTTGTAGACGAGCTGGAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609346 | ||||||||||||||||||||
Literature Links: |
ADAMTSL5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADAMTSL5 - ADAMTS like 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_213604.2 | 1509 | Missense Mutation | TCG,TTG | S,L 401 | NP_998769.2 | |
XM_005259549.3 | 1509 | Missense Mutation | TCG,TTG | S,L 411 | XP_005259606.1 | |
XM_011527961.1 | 1509 | Missense Mutation | TCG,TTG | S,L 415 | XP_011526263.1 | |
XM_011527963.1 | 1509 | Missense Mutation | TCG,TTG | S,L 170 | XP_011526265.1 | |
XM_017026744.1 | 1509 | Missense Mutation | TCG,TTG | S,L 405 | XP_016882233.1 | |
XM_017026745.1 | 1509 | Missense Mutation | TCG,TTG | S,L 401 | XP_016882234.1 |
REEP6 - receptor accessory protein 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |