Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCACAAAGAAGGCCGTGTGGCAGG[G/T]GAACTCCACCACCTTCCTCTGCTCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 182350 MIM: 604925 | ||||||||||||||||||||
Literature Links: |
ATP1A3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ATP1A3 - ATPase Na+/K+ transporting subunit alpha 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256213.1 | 2958 | Missense Mutation | ACC,CCC | T,P 918 | NP_001243142.1 | |
NM_001256214.1 | 2958 | Missense Mutation | ACC,CCC | T,P 920 | NP_001243143.1 | |
NM_152296.4 | 2958 | Missense Mutation | ACC,CCC | T,P 907 | NP_689509.1 |
RABAC1 - Rab acceptor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |