Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTCAAGGGAGACTTCTGCCAGCT[C/T]GCTTCTGCTCTGCTGATGGCCTCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
HHIPL2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HHIPL2 - HHIP like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024746.3 | 2213 | Missense Mutation | NP_079022.2 | |||
XM_011509985.2 | 2213 | Missense Mutation | XP_011508287.1 | |||
XM_011509986.1 | 2213 | Intron | XP_011508288.1 | |||
XM_017002350.1 | 2213 | Intron | XP_016857839.1 |