Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAATAACTTTCATCCACAAAGAAG[A/C]ACCCCCTGAAATTATTAGTCAAGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF501 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF501 - zinc finger protein 501 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF502 - zinc finger protein 502 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134440.1 | 1775 | Missense Mutation | GAA,GCA | E,A 93 | NP_001127912.1 | |
NM_001134441.1 | 1775 | Missense Mutation | GAA,GCA | E,A 93 | NP_001127913.1 | |
NM_001134442.1 | 1775 | Missense Mutation | GAA,GCA | E,A 93 | NP_001127914.1 | |
NM_001282880.1 | 1775 | Missense Mutation | GAA,GCA | E,A 93 | NP_001269809.1 | |
NM_033210.4 | 1775 | Missense Mutation | GAA,GCA | E,A 93 | NP_149987.2 | |
XM_017007440.1 | 1775 | Missense Mutation | GAA,GCA | E,A 93 | XP_016862929.1 |