Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACCGAAGTCATAGTTGACCCGCAC[A/G]TTGGGCAGAGGGGGCACGTACGGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609948 | ||||||||||||||||||||
Literature Links: |
LOC107986762 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC107986762 - uncharacterized LOC107986762 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNF216 - ring finger protein 216 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207111.3 | 2677 | Silent Mutation | AAC,AAT | N,N 893 | NP_996994.1 | |
NM_207116.2 | 2677 | Silent Mutation | AAC,AAT | N,N 836 | NP_996999.1 | |
XM_005249785.2 | 2677 | Silent Mutation | AAC,AAT | N,N 893 | XP_005249842.1 | |
XM_011515436.1 | 2677 | Silent Mutation | AAC,AAT | N,N 458 | XP_011513738.1 | |
XM_017012363.1 | 2677 | Silent Mutation | AAC,AAT | N,N 836 | XP_016867852.1 | |
XM_017012364.1 | 2677 | Intron | XP_016867853.1 | |||
XM_017012365.1 | 2677 | Silent Mutation | AAC,AAT | N,N 458 | XP_016867854.1 |