Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGCCTACTTTCATAGTCTGGCAAA[C/G]GTAGAGTGACAGTTTATGAGTTTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C7orf62 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C7orf62 - chromosome 7 open reading frame 62 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152706.3 | 735 | Missense Mutation | CTT,GTT | L,V 185 | NP_689919.1 | |
XM_011515904.2 | 735 | Missense Mutation | CTT,GTT | L,V 185 | XP_011514206.1 |
ZNF804B - zinc finger protein 804B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181646.3 | 735 | Intron | NP_857597.1 |