Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609045 | ||||||||||||||||||||
Literature Links: |
GPR141 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GPR141 - G protein-coupled receptor 141 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011515374.2 | Intron | XP_011513676.1 | ||||
XM_011515375.2 | Intron | XP_011513677.1 | ||||
XM_011515383.2 | Intron | XP_011513685.1 | ||||
XM_011515385.2 | Intron | XP_011513687.1 | ||||
XM_011515386.2 | Intron | XP_011513688.1 | ||||
XM_011515388.2 | Intron | XP_011513690.1 | ||||
XM_011515389.2 | Intron | XP_011513691.1 | ||||
XM_017012160.1 | Intron | XP_016867649.1 | ||||
XM_017012161.1 | Intron | XP_016867650.1 | ||||
XM_017012162.1 | Intron | XP_016867651.1 | ||||
XM_017012163.1 | Intron | XP_016867652.1 | ||||
XM_017012164.1 | Intron | XP_016867653.1 | ||||
XM_017012165.1 | Intron | XP_016867654.1 | ||||
XM_017012166.1 | Intron | XP_016867655.1 | ||||
XM_017012167.1 | Intron | XP_016867656.1 |