Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606555 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRIM9 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TRIM9 - tripartite motif containing 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015163.5 | 4854 | UTR 3 | NP_055978.4 | |||
NM_052978.4 | 4854 | Intron | NP_443210.1 | |||
XM_005267301.3 | 4854 | UTR 3 | XP_005267358.1 | |||
XM_005267302.3 | 4854 | UTR 3 | XP_005267359.1 | |||
XM_006720018.3 | 4854 | UTR 3 | XP_006720081.1 | |||
XM_011536389.2 | 4854 | Intron | XP_011534691.1 | |||
XM_017020943.1 | 4854 | UTR 3 | XP_016876432.1 | |||
XM_017020944.1 | 4854 | UTR 3 | XP_016876433.1 | |||
XM_017020945.1 | 4854 | UTR 3 | XP_016876434.1 | |||
XM_017020946.1 | 4854 | UTR 3 | XP_016876435.1 | |||
XM_017020947.1 | 4854 | UTR 3 | XP_016876436.1 | |||
XM_017020948.1 | 4854 | UTR 3 | XP_016876437.1 | |||
XM_017020949.1 | 4854 | UTR 3 | XP_016876438.1 | |||
XM_017020950.1 | 4854 | UTR 3 | XP_016876439.1 |
Set Membership: |
HapMap |