Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608684 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NIN PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
NIN - ninein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016350.4 | 6246 | Silent Mutation | CCC,CCT | P,P 1355 | NP_057434.4 | |
NM_020921.3 | 6246 | Intron | NP_065972.3 | |||
NM_182944.2 | 6246 | Intron | NP_891989.2 | |||
NM_182946.1 | 6246 | Silent Mutation | CCC,CCT | P,P 2068 | NP_891991.1 | |
XM_011536817.1 | 6246 | Intron | XP_011535119.1 | |||
XM_011536819.2 | 6246 | Intron | XP_011535121.1 | |||
XM_011536822.1 | 6246 | Intron | XP_011535124.1 | |||
XM_011536823.2 | 6246 | Silent Mutation | CCC,CCT | P,P 2074 | XP_011535125.1 |