Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
VSTM4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
VSTM4 - V-set and transmembrane domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031746.4 | 3371 | UTR 3 | NP_001026916.2 | |||
NM_144984.3 | 3371 | Intron | NP_659421.1 | |||
XM_017015825.1 | 3371 | Intron | XP_016871314.1 | |||
XM_017015826.1 | 3371 | Intron | XP_016871315.1 | |||
XM_017015827.1 | 3371 | Intron | XP_016871316.1 | |||
XM_017015828.1 | 3371 | UTR 3 | XP_016871317.1 |
Set Membership: |
HapMap |