Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 100720 MIM: 613858 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHRND PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CHRND - cholinergic receptor nicotinic delta subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000751.2 | 40 | Silent Mutation | CCA,CCG | P,P 4 | NP_000742.1 | |
NM_001256657.1 | 40 | Silent Mutation | CCA,CCG | P,P 4 | NP_001243586.1 | |
NM_001311195.1 | 40 | UTR 5 | NP_001298124.1 | |||
NM_001311196.1 | 40 | UTR 5 | NP_001298125.1 | |||
XM_011510524.2 | 40 | UTR 5 | XP_011508826.1 |
PRSS56 - protease, serine 56 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195129.1 | 40 | Intron | NP_001182058.1 |
Set Membership: |
HapMap Validated |