Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACAGCCACAGAAGAAGCGGGAGCC[A/G]TAGCCATGACCATAGCCACAGCAGG
Species: |
Human |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
KRTAP22-1 PubMed Links |
Allele Nomenclature: |
|
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KRTAP22-1 - keratin associated protein 22-1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KRTAP22-2 - keratin associated protein 22-2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KRTAP6-2 - keratin associated protein 6-2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181604.1 | 132 | Silent Mutation | TAC,TAT | Y,Y 44 | NP_853635.1 |
KRTAP6-3 - keratin associated protein 6-3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |