Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCTTTGCCGCAGGGGCGCGGGGC[C/T]GAGGGATGTCCGGGGGTTGTGGGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604226 MIM: 601982 MIM: 602945 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARPC4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ARPC4 - actin related protein 2/3 complex subunit 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024959.2 | Intron | NP_001020130.1 | ||||
NM_001024960.2 | Intron | NP_001020131.1 | ||||
NM_001198780.1 | Intron | NP_001185709.1 | ||||
NM_005718.4 | Intron | NP_005709.1 |
ARPC4-TTLL3 - ARPC4-TTLL3 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198793.1 | Intron | NP_001185722.1 |
OGG1 - 8-oxoguanine DNA glycosylase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TADA3 - transcriptional adaptor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278270.1 | Intron | NP_001265199.1 | ||||
NM_006354.3 | Intron | NP_006345.1 | ||||
NM_133480.2 | Intron | NP_597814.1 |
Set Membership: |
HapMap |