Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCCAGCTCCTCCCAGCTGGAAGAC[A/G]CTAGGGTGAACGGAGATAAAAGCAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DEUP1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DEUP1 - deuterosome assembly protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181645.3 | 1090 | Intron | NP_857596.2 | |||
XM_005273802.1 | 1090 | Intron | XP_005273859.1 | |||
XM_011542629.2 | 1090 | Intron | XP_011540931.1 | |||
XM_011542630.1 | 1090 | Intron | XP_011540932.1 | |||
XM_011542631.2 | 1090 | UTR 5 | XP_011540933.1 | |||
XM_011542632.2 | 1090 | Intron | XP_011540934.1 | |||
XM_011542633.2 | 1090 | Intron | XP_011540935.1 | |||
XM_011542635.2 | 1090 | Intron | XP_011540937.1 | |||
XM_011542636.2 | 1090 | Intron | XP_011540938.1 | |||
XM_011542637.2 | 1090 | Intron | XP_011540939.1 | |||
XM_011542638.2 | 1090 | Intron | XP_011540940.1 | |||
XM_017017245.1 | 1090 | Intron | XP_016872734.1 |
LOC102723746 - uncharacterized LOC102723746 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |