Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606211 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRD24 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ANKRD24 - ankyrin repeat domain 24 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_133475.1 | 202 | UTR 5 | NP_597732.1 | |||
XM_011527756.2 | 202 | Missense Mutation | TGG,TGT | W,C 27 | XP_011526058.1 | |
XM_011527757.2 | 202 | Missense Mutation | TGG,TGT | W,C 82 | XP_011526059.1 | |
XM_011527758.2 | 202 | Missense Mutation | TGG,TGT | W,C 27 | XP_011526060.1 | |
XM_011527761.1 | 202 | UTR 5 | XP_011526063.1 | |||
XM_011527764.2 | 202 | Missense Mutation | TGG,TGT | W,C 82 | XP_011526066.1 | |
XM_011527765.2 | 202 | Intron | XP_011526067.1 | |||
XM_011527768.2 | 202 | Intron | XP_011526070.1 | |||
XM_017026438.1 | 202 | UTR 5 | XP_016881927.1 | |||
XM_017026439.1 | 202 | UTR 5 | XP_016881928.1 | |||
XM_017026440.1 | 202 | UTR 5 | XP_016881929.1 | |||
XM_017026441.1 | 202 | Intron | XP_016881930.1 |
SIRT6 - sirtuin 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |