Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
37 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
METTL13 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
METTL13 - methyltransferase like 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007239.1 | 6 | UTR 5 | NP_001007240.1 | |||
NM_014955.2 | 6 | UTR 5 | NP_055770.1 | |||
NM_015935.4 | 6 | UTR 5 | NP_057019.3 | |||
XM_017001455.1 | 6 | Intron | XP_016856944.1 | |||
XM_017001456.1 | 6 | Intron | XP_016856945.1 | |||
XM_017001457.1 | 6 | Intron | XP_016856946.1 | |||
XM_017001458.1 | 6 | UTR 5 | XP_016856947.1 | |||
XM_017001459.1 | 6 | UTR 5 | XP_016856948.1 |
Set Membership: |
HapMap JSNP |