Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGACCACGTGCAATATACGCTGGT[C/T]CCAGTTTCTGGGTGGCAAGAACTTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603465 MIM: 601754 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDC45 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CDC45 - cell division cycle 45 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001178010.2 | 293 | Silent Mutation | GTC,GTT | V,V 50 | NP_001171481.1 | |
NM_001178011.2 | 293 | Silent Mutation | GTC,GTT | V,V 50 | NP_001171482.1 | |
NM_003504.4 | 293 | Silent Mutation | GTC,GTT | V,V 50 | NP_003495.1 | |
XM_005261285.2 | 293 | Silent Mutation | GTC,GTT | V,V 50 | XP_005261342.1 | |
XM_005261286.3 | 293 | Silent Mutation | GTC,GTT | V,V 50 | XP_005261343.1 | |
XM_011530415.1 | 293 | Silent Mutation | GTC,GTT | V,V 50 | XP_011528717.1 | |
XM_011530416.1 | 293 | Silent Mutation | GTC,GTT | V,V 38 | XP_011528718.1 | |
XM_011530417.2 | 293 | Silent Mutation | GTC,GTT | V,V 50 | XP_011528719.1 | |
XM_011530418.2 | 293 | Silent Mutation | GTC,GTT | V,V 50 | XP_011528720.1 | |
XM_017028966.1 | 293 | Intron | XP_016884455.1 | |||
XM_017028967.1 | 293 | Intron | XP_016884456.1 |
UFD1L - ubiquitin fusion degradation 1 like (yeast) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |