Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTGTCGGGTGGAAAGAGGCCCCC[C/G]CTCCTCGCCCCAGCAGTGGCAGTCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612428 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100291105 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC100291105 - uncharacterized LOC100291105 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RBM38 - RNA binding motif protein 38 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291780.1 | Intron | NP_001278709.1 | ||||
NM_017495.5 | Intron | NP_059965.2 | ||||
NM_183425.2 | Intron | NP_906270.1 | ||||
XM_005260446.3 | Intron | XP_005260503.1 | ||||
XM_011528885.2 | Intron | XP_011527187.1 |