Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTCAAGTTATTTGTCCAGGTCTA[C/T]CATAGCTACTGAGCAAATGGGTTGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601951 MIM: 615811 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLK1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CLK1 - CDC like kinase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPIL3 - peptidylprolyl isomerase like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032472.3 | 755 | UTR 3 | NP_115861.1 | |||
NM_130906.2 | 755 | UTR 3 | NP_570981.1 | |||
XM_005246651.3 | 755 | UTR 3 | XP_005246708.1 | |||
XM_005246652.4 | 755 | UTR 3 | XP_005246709.1 | |||
XM_011511358.2 | 755 | UTR 3 | XP_011509660.1 | |||
XM_017004352.1 | 755 | UTR 3 | XP_016859841.1 | |||
XM_017004353.1 | 755 | UTR 3 | XP_016859842.1 | |||
XM_017004354.1 | 755 | Intron | XP_016859843.1 | |||
XM_017004355.1 | 755 | Intron | XP_016859844.1 | |||
XM_017004356.1 | 755 | Intron | XP_016859845.1 | |||
XM_017004357.1 | 755 | UTR 3 | XP_016859846.1 | |||
XM_017004358.1 | 755 | UTR 3 | XP_016859847.1 | |||
XM_017004359.1 | 755 | UTR 3 | XP_016859848.1 | |||
XM_017004360.1 | 755 | UTR 3 | XP_016859849.1 |
Set Membership: |
HapMap |