Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGCCCATGACCATCTCTGCATCT[A/G]GGCTTGGAGGAAGAGGTGGCCCATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608305 | ||||||||||||||||||||
Literature Links: |
SLC13A5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC13A5 - solute carrier family 13 member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143838.2 | 1827 | UTR 3 | NP_001137310.1 | |||
NM_001284509.1 | 1827 | UTR 3 | NP_001271438.1 | |||
NM_001284510.1 | 1827 | UTR 3 | NP_001271439.1 | |||
NM_177550.4 | 1827 | UTR 3 | NP_808218.1 |