Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCTCTTTGATGTTTTTATAGGAT[A/G]TAATGAAAGATGAATGCTCGATGCT
Species: |
Human |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
CCSER1 PubMed Links |
Allele Nomenclature: |
|
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCSER1 - coiled-coil serine rich protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145065.1 | 2589 | Missense Mutation | ATA,GTA | I,V 672 | NP_001138537.1 | |
NM_207491.2 | 2589 | Intron | NP_997374.1 | |||
XM_011531936.1 | 2589 | Missense Mutation | ATA,GTA | I,V 704 | XP_011530238.1 | |
XM_011531937.2 | 2589 | Missense Mutation | ATA,GTA | I,V 704 | XP_011530239.1 | |
XM_011531938.2 | 2589 | Missense Mutation | ATA,GTA | I,V 704 | XP_011530240.1 | |
XM_011531939.2 | 2589 | Missense Mutation | ATA,GTA | I,V 704 | XP_011530241.1 | |
XM_011531941.2 | 2589 | Missense Mutation | ATA,GTA | I,V 704 | XP_011530243.1 | |
XM_011531942.2 | 2589 | Missense Mutation | ATA,GTA | I,V 704 | XP_011530244.1 | |
XM_011531943.2 | 2589 | Missense Mutation | ATA,GTA | I,V 704 | XP_011530245.1 | |
XM_011531945.1 | 2589 | Missense Mutation | ATA,GTA | I,V 704 | XP_011530247.1 | |
XM_011531946.2 | 2589 | Missense Mutation | ATA,GTA | I,V 704 | XP_011530248.1 | |
XM_011531947.2 | 2589 | Missense Mutation | ATA,GTA | I,V 704 | XP_011530249.1 | |
XM_011531948.1 | 2589 | Missense Mutation | ATA,GTA | I,V 704 | XP_011530250.1 | |
XM_011531949.2 | 2589 | Missense Mutation | ATA,GTA | I,V 704 | XP_011530251.1 | |
XM_011531950.2 | 2589 | Missense Mutation | ATA,GTA | I,V 704 | XP_011530252.1 | |
XM_011531951.2 | 2589 | Missense Mutation | ATA,GTA | I,V 704 | XP_011530253.1 | |
XM_011531955.2 | 2589 | Intron | XP_011530257.1 | |||
XM_011531956.2 | 2589 | Missense Mutation | ATA,GTA | I,V 704 | XP_011530258.1 | |
XM_011531957.2 | 2589 | Intron | XP_011530259.1 | |||
XM_011531958.2 | 2589 | Intron | XP_011530260.1 | |||
XM_017008194.1 | 2589 | Missense Mutation | ATA,GTA | I,V 704 | XP_016863683.1 | |
XM_017008195.1 | 2589 | Missense Mutation | ATA,GTA | I,V 704 | XP_016863684.1 | |
XM_017008196.1 | 2589 | Intron | XP_016863685.1 | |||
XM_017008197.1 | 2589 | Missense Mutation | ATA,GTA | I,V 672 | XP_016863686.1 | |
XM_017008198.1 | 2589 | Intron | XP_016863687.1 | |||
XM_017008199.1 | 2589 | Missense Mutation | ATA,GTA | I,V 672 | XP_016863688.1 | |
XM_017008200.1 | 2589 | Intron | XP_016863689.1 | |||
XM_017008201.1 | 2589 | Intron | XP_016863690.1 | |||
XM_017008202.1 | 2589 | Intron | XP_016863691.1 |