Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCGATGGGCTCCGTGCTGAGCAC[C/T]GACAGCGGCAAATCGGCGCCCGCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610432 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNF125 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RNF125 - ring finger protein 125 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017831.3 | 672 | Silent Mutation | ACC,ACT | T,T 7 | NP_060301.2 | |
XM_011526045.2 | 672 | Silent Mutation | ACC,ACT | T,T 7 | XP_011524347.1 | |
XM_011526046.2 | 672 | Silent Mutation | ACC,ACT | T,T 7 | XP_011524348.1 | |
XM_011526047.2 | 672 | Silent Mutation | ACC,ACT | T,T 7 | XP_011524349.1 |