Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600190 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TLE3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TLE3 - transducin like enhancer of split 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105192.2 | Intron | NP_001098662.1 | ||||
NM_001282979.1 | Intron | NP_001269908.1 | ||||
NM_001282980.1 | Intron | NP_001269909.1 | ||||
NM_001282981.1 | Intron | NP_001269910.1 | ||||
NM_001282982.1 | Intron | NP_001269911.1 | ||||
NM_005078.3 | Intron | NP_005069.2 | ||||
NM_020908.2 | Intron | NP_065959.1 | ||||
XM_005254622.4 | Intron | XP_005254679.1 | ||||
XM_005254623.3 | Intron | XP_005254680.1 | ||||
XM_005254625.3 | Intron | XP_005254682.1 | ||||
XM_005254628.3 | Intron | XP_005254685.1 | ||||
XM_005254633.2 | Intron | XP_005254690.1 | ||||
XM_006720665.3 | Intron | XP_006720728.1 | ||||
XM_011521976.2 | Intron | XP_011520278.1 | ||||
XM_011521977.2 | Intron | XP_011520279.1 | ||||
XM_011521978.2 | Intron | XP_011520280.1 | ||||
XM_011521979.2 | Intron | XP_011520281.1 | ||||
XM_011521980.2 | Intron | XP_011520282.1 | ||||
XM_011521981.1 | Intron | XP_011520283.1 | ||||
XM_011521982.2 | Intron | XP_011520284.1 | ||||
XM_011521983.2 | Intron | XP_011520285.1 | ||||
XM_017022532.1 | Intron | XP_016878021.1 |
Set Membership: |
HapMap |