Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601057 MIM: 600857 | ||||||||||||||||||||
Literature Links: |
HRAT5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HRAT5 - heart tissue-associated transcript 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PDCD6 - programmed cell death 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001267556.1 | Intron | NP_001254485.1 | ||||
NM_001267557.1 | Intron | NP_001254486.1 | ||||
NM_001267558.1 | Intron | NP_001254487.1 | ||||
NM_001267559.1 | Intron | NP_001254488.1 | ||||
NM_013232.3 | Intron | NP_037364.1 |
SDHA - succinate dehydrogenase complex flavoprotein subunit A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |