Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602992 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LAIR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LAIR1 - leukocyte associated immunoglobulin like receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289023.2 | 1956 | UTR 3 | NP_001275952.2 | |||
NM_001289025.2 | 1956 | UTR 3 | NP_001275954.2 | |||
NM_001289026.2 | 1956 | UTR 3 | NP_001275955.2 | |||
NM_001289027.2 | 1956 | UTR 3 | NP_001275956.2 | |||
NM_002287.5 | 1956 | UTR 3 | NP_002278.2 | |||
NM_021706.4 | 1956 | UTR 3 | NP_068352.2 | |||
XM_017026803.1 | 1956 | Intron | XP_016882292.1 |