Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTACTTGGGTTATTATCGCTGATTA[C/T]AGCTGGAAACAATTGATTTGCTCTT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 608004 | |||||||||||||||||||||||
Literature Links: |
NFKBIZ PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
NFKBIZ - NFKB inhibitor zeta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005474.2 | 135 | UTR 5 | NP_001005474.1 | |||
NM_031419.3 | 135 | Intron | NP_113607.1 |
NXPE3 - neurexophilin and PC-esterase domain family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134456.1 | 135 | UTR 3 | NP_001127928.1 | |||
NM_145037.2 | 135 | UTR 3 | NP_659474.1 |