Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603107 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TCF20 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TCF20 - transcription factor 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005650.3 | Intron | NP_005641.1 | ||||
NM_181492.2 | Intron | NP_852469.1 | ||||
XM_005261722.3 | Intron | XP_005261779.1 | ||||
XM_006724313.3 | Intron | XP_006724376.1 | ||||
XM_011530353.2 | Intron | XP_011528655.1 | ||||
XM_011530354.2 | Intron | XP_011528656.1 |
Set Membership: |
HapMap |