Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTCTCTGGTGCGCTCGCCCCTCC[C/T]CTCTGAGGAGATGTTATTTCCCTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616856 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BRPF3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
BRPF3 - bromodomain and PHD finger containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015695.2 | 20 | Intron | NP_056510.2 | |||
XM_005249010.2 | 20 | Intron | XP_005249067.1 | |||
XM_005249011.3 | 20 | UTR 5 | XP_005249068.1 | |||
XM_011514489.1 | 20 | UTR 5 | XP_011512791.1 | |||
XM_011514490.1 | 20 | Intron | XP_011512792.1 | |||
XM_011514491.1 | 20 | Intron | XP_011512793.1 | |||
XM_011514492.1 | 20 | Intron | XP_011512794.1 | |||
XM_017010742.1 | 20 | Intron | XP_016866231.1 | |||
XM_017010743.1 | 20 | Intron | XP_016866232.1 |
LOC102723648 - uncharacterized LOC102723648 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |