Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607494 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
INPP4B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
INPP4B - inositol polyphosphate-4-phosphatase type II B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001101669.1 | Intron | NP_001095139.1 | ||||
NM_003866.2 | Intron | NP_003857.2 | ||||
XM_005263323.3 | Intron | XP_005263380.1 | ||||
XM_005263325.3 | Intron | XP_005263382.1 | ||||
XM_011532391.2 | Intron | XP_011530693.1 | ||||
XM_011532392.2 | Intron | XP_011530694.1 | ||||
XM_017008794.1 | Intron | XP_016864283.1 | ||||
XM_017008795.1 | Intron | XP_016864284.1 | ||||
XM_017008796.1 | Intron | XP_016864285.1 | ||||
XM_017008797.1 | Intron | XP_016864286.1 | ||||
XM_017008798.1 | Intron | XP_016864287.1 |