Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607247 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHODL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHODL - chondrolectin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204174.1 | Intron | NP_001191103.1 | ||||
NM_001204175.1 | Intron | NP_001191104.1 | ||||
NM_001204176.1 | Intron | NP_001191105.1 | ||||
NM_001204177.1 | Intron | NP_001191106.1 | ||||
NM_001204178.1 | Intron | NP_001191107.1 | ||||
NM_024944.2 | Intron | NP_079220.2 | ||||
XM_011529453.2 | Intron | XP_011527755.1 | ||||
XM_011529457.2 | Intron | XP_011527759.1 | ||||
XM_017028273.1 | Intron | XP_016883762.1 |
CHODL-AS1 - CHODL antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |