Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
46 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616690 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEP104 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CEP104 - centrosomal protein 104 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014704.3 | 4968 | UTR 3 | NP_055519.1 | |||
XM_005244815.4 | 4968 | Intron | XP_005244872.1 | |||
XM_011542473.2 | 4968 | UTR 3 | XP_011540775.1 | |||
XM_011542474.2 | 4968 | Intron | XP_011540776.1 | |||
XM_011542475.2 | 4968 | Intron | XP_011540777.1 | |||
XM_011542476.2 | 4968 | Intron | XP_011540778.1 | |||
XM_011542477.2 | 4968 | Intron | XP_011540779.1 | |||
XM_011542478.1 | 4968 | Intron | XP_011540780.1 | |||
XM_017002917.1 | 4968 | Intron | XP_016858406.1 | |||
XM_017002918.1 | 4968 | Intron | XP_016858407.1 | |||
XM_017002919.1 | 4968 | Intron | XP_016858408.1 |