Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGATTAGGTGGGCCTGTAAAAGAT[A/T]TCGGCAAAAAAAAAAAAAAAAAAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 159558 | ||||||||||||||||||||
Literature Links: |
MLLT3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MLLT3 - MLLT3, super elongation complex subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286691.1 | 5026 | UTR 3 | NP_001273620.1 | |||
NM_004529.3 | 5026 | UTR 3 | NP_004520.2 | |||
XM_017014726.1 | 5026 | UTR 3 | XP_016870215.1 | |||
XM_017014727.1 | 5026 | UTR 3 | XP_016870216.1 |