Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTCTTTTTAAAATTTGTTACTTG[G/T]TTATTTATCAAGAGAAACTATTTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609031 | ||||||||||||||||||||
Literature Links: |
EPPIN PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EPPIN - epididymal peptidase inhibitor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
EPPIN-WFDC6 - EPPIN-WFDC6 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198986.1 | 1455 | UTR 3 | NP_001185915.1 |
WFDC6 - WAP four-disulfide core domain 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080827.1 | 1455 | Intron | NP_543017.1 |