Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGCCACTTTTATTTTTATTTAGA[A/G]TTTTTTTTAGAGACAGGGTTTGGCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607698 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LCOR PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
LCOR - ligand dependent nuclear receptor corepressor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170765.1 | Intron | NP_001164236.1 | ||||
NM_001170766.1 | Intron | NP_001164237.1 | ||||
NM_032440.3 | Intron | NP_115816.2 | ||||
XM_006718033.3 | Intron | XP_006718096.1 | ||||
XM_011540277.2 | Intron | XP_011538579.1 | ||||
XM_017016784.1 | Intron | XP_016872273.1 | ||||
XM_017016785.1 | Intron | XP_016872274.1 | ||||
XM_017016786.1 | Intron | XP_016872275.1 | ||||
XM_017016787.1 | Intron | XP_016872276.1 | ||||
XM_017016788.1 | Intron | XP_016872277.1 |
LOC102723665 - uncharacterized LOC102723665 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR607 - microRNA 607 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |