Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAACTCTATAGCCCATACCTCACC[C/T]TTGTCTCAGCATCTGTCTCTTCCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602332 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NCAPH PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NCAPH - non-SMC condensin I complex subunit H | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281710.1 | Intron | NP_001268639.1 | ||||
NM_001281711.1 | Intron | NP_001268640.1 | ||||
NM_001281712.1 | Intron | NP_001268641.1 | ||||
NM_015341.4 | Intron | NP_056156.2 | ||||
XM_005263908.3 | Intron | XP_005263965.1 | ||||
XM_006712388.3 | Intron | XP_006712451.1 |