Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611569 MIM: 610903 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CKAP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CKAP2 - cytoskeleton associated protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098525.2 | Intron | NP_001091995.1 | ||||
NM_001286686.1 | Intron | NP_001273615.1 | ||||
NM_001286687.1 | Intron | NP_001273616.1 | ||||
NM_018204.4 | Intron | NP_060674.3 | ||||
XM_005266344.4 | Intron | XP_005266401.1 | ||||
XM_011535043.2 | Intron | XP_011533345.1 |
VPS36 - vacuolar protein sorting 36 homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |