Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTCAGTAAATCCAAGGTCCTTCA[G/T]GACATGTAGGAGAAGTACAAGCACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 187410 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNTT PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DNTT - DNA nucleotidylexotransferase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OPALIN - oligodendrocytic myelin paranodal and inner loop protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040102.2 | 2927 | UTR 3 | NP_001035191.1 | |||
NM_001040103.2 | 2927 | UTR 3 | NP_001035192.1 | |||
NM_001284320.1 | 2927 | UTR 3 | NP_001271249.1 | |||
NM_001284321.1 | 2927 | UTR 3 | NP_001271250.1 | |||
NM_001284322.1 | 2927 | UTR 3 | NP_001271251.1 | |||
NM_001284323.1 | 2927 | UTR 3 | NP_001271252.1 | |||
NM_001284324.1 | 2927 | UTR 3 | NP_001271253.1 | |||
NM_001284326.1 | 2927 | UTR 3 | NP_001271255.1 | |||
NM_001284327.1 | 2927 | UTR 3 | NP_001271256.1 | |||
NM_033207.4 | 2927 | UTR 3 | NP_149984.1 |