Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCAGGTAGGCAGAGATGGCAGTGC[G/T]CTGATTCTGCTGGTGGACTGAAGTC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611595 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HSBP1L1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HSBP1L1 - heat shock factor binding protein 1 like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136180.1 | Intron | NP_001129652.1 |
TXNL4A - thioredoxin like 4A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |