Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAAAACCATGACCAGTATCCTTTA[A/G]AAGAATGTTATGCTGTCTTCATATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
WDYHV1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
WDYHV1 - WDYHV motif containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001283024.1 | 191 | UTR 5 | NP_001269953.1 | |||
NM_001283027.1 | 191 | UTR 5 | NP_001269956.1 | |||
NM_018024.2 | 191 | Missense Mutation | AAA,GAA | K,E 48 | NP_060494.1 | |
XM_006716597.3 | 191 | Missense Mutation | AAA,GAA | K,E 48 | XP_006716660.1 | |
XM_011517147.1 | 191 | Missense Mutation | AAA,GAA | K,E 48 | XP_011515449.1 | |
XM_011517151.2 | 191 | UTR 5 | XP_011515453.1 | |||
XM_017013601.1 | 191 | Missense Mutation | AAA,GAA | K,E 48 | XP_016869090.1 | |
XM_017013602.1 | 191 | UTR 5 | XP_016869091.1 | |||
XM_017013603.1 | 191 | UTR 5 | XP_016869092.1 |