Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGCAACCAGAGCGGCGGCGGGAA[A/G]TGTTGCGCAGGCGCATCCGATCGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
46 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603311 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDC7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CDC7 - cell division cycle 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134419.1 | 3 | UTR 5 | NP_001127891.1 | |||
NM_001134420.1 | 3 | Intron | NP_001127892.1 | |||
NM_003503.3 | 3 | UTR 5 | NP_003494.1 | |||
XM_005271241.2 | 3 | UTR 5 | XP_005271298.1 | |||
XM_005271244.2 | 3 | UTR 5 | XP_005271301.1 | |||
XM_017002425.1 | 3 | UTR 5 | XP_016857914.1 | |||
XM_017002426.1 | 3 | Intron | XP_016857915.1 | |||
XM_017002427.1 | 3 | Intron | XP_016857916.1 |
Set Membership: |
HapMap |