Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGTTGGAACCAGACAAAAGCCTT[C/T]CTGTGCAAAAATGTAGGAGTGTGAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138253 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GRIN2A PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GRIN2A - glutamate ionotropic receptor NMDA type subunit 2A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000833.4 | 10133 | UTR 3 | NP_000824.1 | |||
NM_001134407.2 | 10133 | UTR 3 | NP_001127879.1 | |||
NM_001134408.2 | 10133 | UTR 3 | NP_001127880.1 | |||
XM_011522458.2 | 10133 | UTR 3 | XP_011520760.1 | |||
XM_011522461.2 | 10133 | UTR 3 | XP_011520763.1 | |||
XM_017023172.1 | 10133 | UTR 3 | XP_016878661.1 | |||
XM_017023173.1 | 10133 | UTR 3 | XP_016878662.1 |
Set Membership: |
HapMap |